Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291076.1 | NP_001278005.1 | 1 - 2 | 137 | 159 | |
NM_001291077.1 | NP_001278006.1 | 1 - 2 | 137 | 159 | ||
NM_198214.3 | NP_937857.1 | 1 - 2 | 137 | 159 | ||
GenBank mRNA | AK032279.1 | - | 1 - 2 | 78 | 159 | |
AK047568.1 | - | 1 - 2 | 118 | 159 | ||
AK122262.1 | - | 1 - 2 | 77 | 159 | ||
BC072567.1 | - | 1 - 2 | 100 | 159 | ||
BC096541.1 | - | 1 - 2 | 29 | 159 | ||
BC117896.1 | - | 1 - 2 | 63 | 40096951 | 159 | |
BC117897.1 | - | 1 - 2 | 63 | 40096952 | 159 | |
BC139792.1 | - | 1 - 2 | 63 | 159 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |