Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128170.2 | NP_001121642.1 | 3 - 4 | 1023 | 115 | |
NM_001128171.2 | NP_001121643.1 | 4 - 5 | 1157 | 115 | ||
NM_001276279.1 | NP_001263208.1 | 4 - 5 | 1154 | 115 | ||
NM_173369.3 | NP_775545.1 | 3 - 4 | 1023 | 115 | ||
GenBank mRNA | AK039054.1 | - | 4 - 5 | 1152 | 115 | |
AK042764.1 | - | 3 - 4 | 990 | 115 | ||
AK122389.1 | - | 4 - 5 | 1138 | 115 | ||
AK133479.1 | - | 3 - 4 | 987 | 115 | ||
AK147204.1 | - | 3 - 4 | 992 | 115 | ||
AK147426.1 | - | 3 - 4 | 987 | 115 | ||
AK167424.1 | - | 4 - 5 | 1118 | 115 | ||
BC042438.1 | - | 3 - 4 | 976 | 3983771 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |