Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163660.1 | NP_001157132.1 | 3 - 4 | 319 | 59 | |
NM_027280.3 | NP_081556.3 | 5 - 6 | 529 | 59 | ||
GenBank mRNA | AK082367.1 | - | 5 - 6 | 517 | 59 | |
AK166564.1 | - | 5 - 6 | 532 | 59 | ||
BC034838.1 | - | 5 - 6 | 503 | 1429943 | 59 | |
DV662736.1 | - | 3 - 4 | 299 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |