Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009954.3 | NP_034084.2 | 1 - 2 | 164 | 58 | |
GenBank mRNA | AK090315.1 | - | 1 - 2 | 165 | 58 | |
AK167516.1 | - | 1 - 2 | 164 | 58 | ||
AK167717.1 | - | 1 - 2 | 165 | 58 | ||
BC057578.1 | - | 1 - 2 | 124 | 6401186 | 58 | |
BY733553.1 | - | 1 - 2 | 165 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |