Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037665.2 | NP_001032754.1 | 1 - 2 | 172 | 56 | |
NM_011742.2 | NP_035872.2 | 1 - 2 | 172 | 56 | ||
GenBank mRNA | AK134452.1 | - | 1 - 2 | 79 | 56 | |
AK147054.1 | - | 1 - 2 | 110 | 56 | ||
AK153830.1 | - | 1 - 2 | 97 | 56 | ||
BC025141.1 | - | 1 - 2 | 135 | 56 | ||
BC055796.1 | - | 1 - 2 | 81 | 6412519 | 56 | |
BC062970.1 | - | 1 - 2 | 81 | 6412519 | 56 | |
X16493.1 | - | 1 - 2 | 281 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |