Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007586.1 | NP_031612.1 | 8 - 9 | 641 | 136 | |
GenBank mRNA | AK032171.1 | - | 647 | 136 | ||
BC017646.1 | - | 8 - 9 | 641 | 4527074 | 136 | |
X73985.1 | - | 6 - 7 | 330 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |