Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028805.2 | NP_083081.2 | 2 - 3 | 400 | 73 | |
GenBank mRNA | AK010364.1 | - | 2 - 3 | 331 | 73 | |
AK031630.1 | - | 2 - 3 | 312 | 73 | ||
AK034103.1 | - | 2 - 3 | 320 | 73 | ||
AK049345.1 | - | 2 - 3 | 314 | 73 | ||
AK054173.1 | - | 2 - 3 | 371 | 73 | ||
AK087973.1 | - | 2 - 3 | 313 | 73 | ||
AK136322.1 | - | 2 - 3 | 320 | 73 | ||
BC045200.1 | - | 1 - 2 | 311 | 5322489 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |