Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008187.2 | NP_032213.2 | 2 - 3 | 457 | 62 | |
GenBank mRNA | AK088713.1 | - | 2 - 3 | 361 | 62 | |
AK172603.1 | - | 2 - 3 | 433 | 62 | ||
BC119023.1 | - | 2 - 3 | 353 | 8733772 | 62 | |
BC119025.1 | - | 2 - 3 | 353 | 8733774 | 62 | |
BM934726.1 | - | 2 - 3 | 457 | 62 | ||
Z54179.1 | - | 2 - 3 | 380 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |