Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100017572.mm.1 |
NM_001102615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017572.mm.1 |
NM_001102615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017572.mm.1 |
TC1100001878.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017573.mm.1 |
NM_001102615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017573.mm.1 |
NM_001102615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017573.mm.1 |
TC1100001878.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017574.mm.1 |
NM_001102615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017574.mm.1 |
NM_001102615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017574.mm.1 |
TC1100001878.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017569.mm.1 |
NM_001102615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017569.mm.1 |
NM_001102615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017569.mm.1 |
TC1100001878.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017571.mm.1 |
NM_001102615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017571.mm.1 |
NM_001102615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017571.mm.1 |
TC1100001878.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100017570.mm.1 |
NM_001102615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017570.mm.1 |
NM_001102615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100017570.mm.1 |
TC1100001878.mm.1 |
MTA TC |