Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102615.1 | NP_001096085.1 | 3 - 4 | 355 | 58 | |
GenBank mRNA | AK155544.1 | - | 3 - 4 | 318 | 58 | |
EF570784.1 | - | 3 - 4 | 230 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |