Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013581.3 | NP_038609.3 | 10 - 11 | 2530 | 56 | |
GenBank mRNA | AF109377.2 | - | 10 - 11 | 2513 | 56 | |
AK031510.1 | - | 9 - 10 | 1948 | 56 | ||
AK034636.2 | - | 10 - 11 | 2535 | 56 | ||
AK152464.1 | - | 10 - 11 | 2521 | 56 | ||
AK158507.1 | - | 10 - 11 | 2892 | 56 | ||
AK159497.1 | - | 10 - 11 | 2532 | 56 | ||
AK162971.1 | - | 10 - 11 | 2531 | 56 | ||
AK171574.1 | - | 10 - 11 | 2524 | 56 | ||
AK220387.1 | - | 9 - 10 | 2974 | 56 | ||
BC049541.1 | - | 4 - 5 | 980 | 56 | ||
BC063056.1 | - | 10 - 11 | 2512 | 5720460 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |