Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164532.1 | NP_001158004.1 | 4 - 5 | 600 | 75 | |
GenBank mRNA | DV648079.1 | - | 4 - 5 | 571 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |