Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emilin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291145.1 | NP_001278074.1 | 1 - 2 | 334 | 104 | |
NM_182840.2 | NP_878260.1 | 1 - 2 | 334 | 104 | ||
GenBank mRNA | BC054738.1 | - | 1 - 2 | 323 | 6402350 | 104 |
BC057619.1 | - | 1 - 2 | 323 | 6402350 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |