Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab40b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139147.3 | NP_631886.2 | 4 - 5 | 449 | 86 | |
GenBank mRNA | AB232638.1 | - | 4 - 5 | 337 | 86 | |
AF425643.1 | - | 4 - 5 | 337 | 86 | ||
BB847804.1 | - | 4 - 5 | 366 | 86 | ||
BC096601.1 | - | 4 - 5 | 443 | 4974190 | 86 | |
BY283181.1 | - | 4 - 5 | 329 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |