Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001333.2 | NP_001001333.2 | 2 - 3 | 546 | 68 | |
NM_001146073.1 | NP_001139545.1 | 1 - 2 | 132 | 68 | ||
GenBank mRNA | AK154237.1 | - | 2 - 3 | 520 | 68 | |
BC056487.1 | - | 2 - 3 | 2127 | 68 | ||
BC069960.1 | - | 1 - 2 | 84 | 5007884 | 68 | |
BY206924.1 | - | 2 - 3 | 234 | 68 | ||
BY296469.1 | - | 3 - 4 | 289 | 68 | ||
FM204885.1 | - | 1 - 2 | 83 | 68 | ||
FM204886.1 | - | 1 - 2 | 83 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |