Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039514.1 | NP_001034603.1 | 5 - 6 | 769 | 147 | |
GenBank mRNA | AK159104.1 | - | 5 - 6 | 770 | 147 | |
BC039963.1 | - | 3 - 4 | 555 | 4240646 | 147 | |
BC061234.1 | - | 4 - 5 | 608 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |