Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153172.1 | NP_694812.1 | 4 - 5 | 604 | 138 | |
GenBank mRNA | AF392062.1 | - | 4 - 5 | 604 | 138 | |
AK077121.1 | - | 4 - 5 | 602 | 138 | ||
BC107230.1 | - | 3 - 4 | 577 | 40052042 | 138 | |
BC107231.1 | - | 3 - 4 | 577 | 40052043 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |