Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083319.1 | NP_001076788.1 | 4 - 5 | 751 | 80 | |
NM_013699.2 | NP_038727.1 | 4 - 5 | 751 | 80 | ||
GenBank mRNA | AK143282.1 | - | 4 - 5 | 752 | 80 | |
AY182062.1 | - | 3 - 4 | 340 | 80 | ||
BC034749.1 | - | 4 - 5 | 440 | 80 | ||
BC139828.1 | - | 3 - 4 | 368 | 40142981 | 80 | |
BC141082.1 | - | 3 - 4 | 430 | 9055694 | 80 | |
U20086.1 | - | 3 - 4 | 342 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |