Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asgr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007493.3 | NP_031519.1 | 1 - 2 | 143 | 75 | |
GenBank mRNA | BC011197.1 | - | 1 - 2 | 143 | 4194949 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |