Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146260.2 | NP_666372.1 | 1 - 2 | 813 | 124 | |
GenBank mRNA | AF481143.1 | - | 1 - 2 | 813 | 124 | |
AK030341.1 | - | 1 - 2 | 843 | 124 | ||
AK031966.1 | - | 1 - 2 | 802 | 124 | ||
BC138840.1 | - | 1 - 2 | 377 | 8861862 | 124 | |
BC141124.1 | - | 1 - 2 | 377 | 9055736 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |