Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146260.2 | NP_666372.1 | 3 - 4 | 1258 | 124 | |
GenBank mRNA | AF481143.1 | - | 3 - 4 | 1258 | 124 | |
AK030341.1 | - | 3 - 4 | 1288 | 124 | ||
BC138840.1 | - | 3 - 4 | 822 | 8861862 | 124 | |
BC141124.1 | - | 3 - 4 | 822 | 9055736 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |