Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026482.2 | NP_080758.1 | 12 - 13 | 2511 | 82 | |
GenBank mRNA | BC029045.1 | - | 6 - 7 | 1151 | 82 | |
BC030901.1 | - | 6 - 7 | 1152 | 82 | ||
BC049262.1 | - | 6 - 7 | 1151 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Mammalian Gene Collection Mus musculus Probe spans exons |