Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026559.3 | NP_080835.1 | 1 - 2 | 219 | 78 | |
GenBank mRNA | AK004219.1 | - | 1 - 2 | 227 | 78 | |
AK007595.1 | - | 1 - 2 | 223 | 78 | ||
AK013103.1 | - | 1 - 2 | 162 | 78 | ||
AK076425.1 | - | 1 - 2 | 219 | 78 | ||
AK169882.1 | - | 1 - 2 | 162 | 78 | ||
BC030344.1 | - | 1 - 2 | 217 | 3673521 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |