Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200018009.mm.1 |
NM_001141975.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018009.mm.1 |
NM_001141975.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018009.mm.1 |
NM_001141976.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018009.mm.1 |
NM_001141976.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018009.mm.1 |
NM_001141977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018009.mm.1 |
NM_001141977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018009.mm.1 |
NM_001141978.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018009.mm.1 |
NM_001141978.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018009.mm.1 |
NM_028109.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018009.mm.1 |
NM_028109.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018009.mm.1 |
TC0200002301.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018012.mm.1 |
NM_001141975.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018012.mm.1 |
NM_001141975.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018012.mm.1 |
NM_001141976.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018012.mm.1 |
NM_001141976.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018012.mm.1 |
NM_001141977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018012.mm.1 |
NM_001141977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018012.mm.1 |
NM_001141978.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018012.mm.1 |
NM_001141978.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018012.mm.1 |
NM_028109.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018012.mm.1 |
NM_028109.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018012.mm.1 |
TC0200002301.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018011.mm.1 |
NM_001141975.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018011.mm.1 |
NM_001141975.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018011.mm.1 |
NM_001141976.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018011.mm.1 |
NM_001141976.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018011.mm.1 |
NM_001141977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018011.mm.1 |
NM_001141977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018011.mm.1 |
NM_001141978.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018011.mm.1 |
NM_001141978.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018011.mm.1 |
NM_028109.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018011.mm.1 |
NM_028109.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018011.mm.1 |
TC0200002301.mm.1 |
MTA TC |