Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001141975.1 | NP_001135447.1 | 5 - 6 | 650 | 84 | |
NM_001141976.1 | NP_001135448.1 | 5 - 6 | 666 | 84 | ||
NM_001141977.1 | NP_001135449.1 | 5 - 6 | 854 | 84 | ||
NM_001141978.1 | NP_001135450.1 | 5 - 6 | 653 | 84 | ||
NM_028109.4 | NP_082385.3 | 5 - 6 | 841 | 84 | ||
GenBank mRNA | AK011311.2 | - | 2 - 3 | 209 | 84 | |
AK090101.1 | - | 5 - 6 | 604 | 84 | ||
AK153957.1 | - | 5 - 6 | 588 | 84 | ||
AK167030.1 | - | 2 - 3 | 306 | 84 | ||
BC060619.1 | - | 5 - 6 | 742 | 6812536 | 84 | |
BU148033.1 | - | 5 - 6 | 568 | 84 | ||
BU612544.1 | - | 1 - 2 | 98 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |