Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008750.5 | NP_032776.1 | 1 - 2 | 443 | 65 | |
GenBank mRNA | AB095441.1 | - | 1 - 2 | 288 | 65 | |
AB095442.1 | - | 1 - 2 | 288 | 65 | ||
AB095443.1 | - | 1 - 2 | 288 | 65 | ||
AB095444.1 | - | 1 - 2 | 288 | 65 | ||
AK012352.1 | - | 1 - 2 | 428 | 65 | ||
AK048109.1 | - | 1 - 2 | 442 | 65 | ||
AK149075.1 | - | 1 - 2 | 427 | 65 | ||
BC004688.1 | - | 1 - 2 | 403 | 3501314 | 65 | |
BC058244.1 | - | 1 - 2 | 443 | 6312910 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |