Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031251.4 | NP_112541.1 | 7 - 8 | 684 | 95 | |
GenBank mRNA | AJ250670.1 | - | 6 - 7 | 582 | 95 | |
AK054467.1 | - | 8 - 9 | 862 | 95 | ||
AK078511.1 | - | 7 - 8 | 696 | 95 | ||
AK079295.1 | - | 7 - 8 | 685 | 95 | ||
AK137143.1 | - | 7 - 8 | 727 | 95 | ||
AK170790.1 | - | 7 - 8 | 696 | 95 | ||
AK170828.1 | - | 7 - 8 | 707 | 95 | ||
BC005479.1 | - | 7 - 8 | 671 | 3484952 | 95 | |
BC006033.1 | - | 7 - 8 | 671 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |