Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankfy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009671.5 | NP_033801.4 | 19 - 20 | 2794 | 93 | |
GenBank mRNA | AB011370.2 | - | 19 - 20 | 2841 | 93 | |
AB098329.1 | - | 19 - 20 | 2792 | 93 | ||
BC139231.1 | - | 19 - 20 | 2805 | 8862253 | 93 | |
BC145583.1 | - | 19 - 20 | 2805 | 9054149 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |