Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubtf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004961.1 | - | 11 - 12 | 1276 | 98 | |
AK038139.1 | - | 7 - 8 | 2406 | 98 | ||
AK128947.1 | - | 11 - 12 | 1303 | 98 | ||
AK158861.1 | - | 11 - 12 | 1275 | 98 | ||
AK158935.1 | - | 11 - 12 | 1234 | 98 | ||
BQ572941.1 | - | 2 - 3 | 147 | 98 | ||
X60831.1 | - | 11 - 12 | 1254 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |