Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161329.1 | NP_001154801.1 | 10 - 11 | 1261 | 117 | |
NM_133702.2 | NP_598463.2 | 10 - 11 | 1261 | 117 | ||
GenBank mRNA | AK005123.1 | - | 10 - 11 | 1150 | 117 | |
AK038696.1 | - | 10 - 11 | 1152 | 117 | ||
AK076704.1 | - | 10 - 11 | 1262 | 117 | ||
AK078524.1 | - | 10 - 11 | 1152 | 117 | ||
AK157743.1 | - | 10 - 11 | 1201 | 117 | ||
BC004785.1 | - | 5 - 6 | 540 | 117 | ||
BC024874.1 | - | 10 - 11 | 1140 | 3984027 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |