Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2o | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173755.3 | NP_776116.2 | 15 - 16 | 2973 | 98 | |
GenBank mRNA | AK045398.1 | - | 2 - 3 | 343 | 98 | |
AK129431.1 | - | 15 - 16 | 2973 | 98 | ||
BC023329.1 | - | 2 - 3 | 703 | 98 | ||
BC039233.1 | - | 7 - 8 | 1439 | 98 | ||
BC051108.1 | - | 6 - 7 | 1318 | 98 | ||
BC055067.1 | - | 1 - 2 | 66 | 97 | ||
BC059193.1 | - | 15 - 16 | 2598 | 4160667 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |