Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204129.1 | NP_001191058.1 | 2 - 3 | 267 | 69 | |
NM_001204130.1 | NP_001191059.1 | 2 - 3 | 179 | 69 | ||
NM_019959.3 | NP_064343.1 | 2 - 3 | 257 | 69 | ||
GenBank mRNA | AF192499.1 | - | 2 - 3 | 205 | 69 | |
AK005484.1 | - | 2 - 3 | 226 | 69 | ||
AK145104.1 | - | 2 - 3 | 180 | 69 | ||
BC037636.1 | - | 1 - 2 | 172 | 69 | ||
BC049894.1 | - | 2 - 3 | 208 | 4018549 | 69 | |
CJ056741.1 | - | 2 - 3 | 257 | 69 | ||
DQ002394.1 | - | 1 - 2 | 151 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |