Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144890.2 | NP_659139.2 | 6 - 7 | 757 | 77 | |
GenBank mRNA | AK142982.1 | - | 6 - 7 | 758 | 77 | |
BC018465.1 | - | 6 - 7 | 719 | 3156626 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |