Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291105.1 | NP_001278034.1 | 1 - 2 | 360 | 62 | |
GenBank mRNA | AK017841.1 | - | 1 - 2 | 352 | 62 | |
AK220226.1 | - | 1 - 2 | 101 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Mus musculus Probe spans exons |