Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blmh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178645.4 | NP_848760.1 | 9 - 10 | 1253 | 61 | |
GenBank mRNA | AK035228.1 | - | 9 - 10 | 1127 | 61 | |
AK040799.1 | - | 9 - 10 | 1143 | 61 | ||
AK049461.1 | - | 9 - 10 | 1166 | 61 | ||
AK051441.1 | - | 9 - 10 | 1152 | 61 | ||
AK053872.1 | - | 9 - 10 | 1133 | 61 | ||
AK150438.1 | - | 9 - 10 | 1152 | 61 | ||
AK150896.1 | - | 9 - 10 | 1133 | 61 | ||
AK167325.1 | - | 9 - 10 | 1133 | 61 | ||
AK219647.1 | - | 2 - 3 | 170 | 62 | ||
BC024090.1 | - | 9 - 10 | 1138 | 5102275 | 61 | |
BC027037.1 | - | 9 - 10 | 1109 | 5364667 | 61 | |
BC027362.1 | - | 9 - 10 | 1142 | 4935208 | 61 | |
BC027403.1 | - | 9 - 10 | 1163 | 4952098 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |