Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vstm2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198627.2 | NP_941029.2 | 1 - 2 | 277 | 68 | |
GenBank mRNA | BC058538.1 | - | 1 - 2 | 165 | 6820223 | 68 |
BF323097.1 | - | 1 - 2 | 277 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |