Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025680.4 | NP_079956.3 | 13 - 14 | 1525 | 81 | |
GenBank mRNA | AK010547.1 | - | 13 - 14 | 1436 | 81 | |
AK044690.1 | - | 13 - 14 | 1465 | 81 | ||
AK077616.1 | - | 13 - 14 | 1469 | 81 | ||
AK145147.1 | - | 13 - 14 | 1481 | 81 | ||
AK145598.1 | - | 13 - 14 | 1469 | 81 | ||
AK145719.1 | - | 13 - 14 | 1469 | 81 | ||
AK145888.1 | - | 13 - 14 | 1486 | 81 | ||
AK146274.1 | - | 13 - 14 | 1485 | 81 | ||
AY009405.1 | - | 13 - 14 | 1485 | 81 | ||
BC038332.1 | - | 3 - 4 | 81 | |||
BC050787.1 | - | 13 - 14 | 1495 | 6705648 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |