Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200019113.mm.1 |
NM_139151.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019113.mm.1 |
NM_139151.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019113.mm.1 |
TC0200002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019121.mm.1 |
NM_139151.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019121.mm.1 |
NM_139151.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019121.mm.1 |
TC0200002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019123.mm.1 |
NM_139151.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019123.mm.1 |
NM_139151.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019123.mm.1 |
TC0200002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019115.mm.1 |
NM_139151.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019115.mm.1 |
NM_139151.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019115.mm.1 |
TC0200002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019114.mm.1 |
NM_139151.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019114.mm.1 |
NM_139151.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019114.mm.1 |
TC0200002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019120.mm.1 |
NM_139151.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019120.mm.1 |
NM_139151.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019120.mm.1 |
TC0200002383.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019126.mm.1 |
NM_139151.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019126.mm.1 |
NM_139151.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019126.mm.1 |
TC0200002383.mm.1 |
MTA TC |