Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029983.5 | NP_084259.1 | 1 - 2 | 200 | 64 | |
GenBank mRNA | AK030877.1 | - | 1 - 2 | 201 | 64 | |
AK138709.1 | - | 1 - 2 | 201 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Mus musculus Probe spans exons |