Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stra13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016665.2 | NP_057874.2 | 3 - 4 | 187 | 84 | |
GenBank mRNA | AK076181.1 | - | 2 - 3 | 246 | 84 | |
AK080500.1 | - | 3 - 4 | 159 | 84 | ||
AK170126.1 | - | 3 - 4 | 159 | 84 | ||
BC052335.1 | - | 4 - 5 | 437 | 84 | ||
CA320983.1 | - | 2 - 3 | 126 | 84 | ||
U95003.1 | - | 4 - 5 | 251 | 84 | ||
U95005.1 | - | 4 - 5 | 425 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |