Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098836.1 | NP_001092306.1 | 10 - 11 | 974 | 90 | |
NM_001098837.1 | NP_001092307.1 | 10 - 11 | 953 | 90 | ||
GenBank mRNA | AK090368.1 | - | 10 - 11 | 955 | 90 | |
BC059880.1 | - | 4 - 5 | 369 | 90 | ||
BC130269.1 | - | 9 - 10 | 724 | 90 | ||
BM950535.1 | - | 8 - 9 | 531 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |