Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178624.6 | NP_848739.1 | 12 - 13 | 944 | 80 | |
GenBank mRNA | AK039010.1 | - | 12 - 13 | 944 | 80 | |
AK044693.1 | - | 12 - 13 | 941 | 80 | ||
AK045742.1 | - | 12 - 13 | 931 | 80 | ||
AK089994.1 | - | 12 - 13 | 920 | 80 | ||
AK141177.1 | - | 12 - 13 | 934 | 80 | ||
AK163472.1 | - | 12 - 13 | 940 | 80 | ||
BC096582.1 | - | 12 - 13 | 930 | 3963599 | 80 | |
BC145665.1 | - | 12 - 13 | 924 | 40130968 | 80 | |
BC145998.1 | - | 12 - 13 | 924 | 40131301 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |