Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21776628 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21776626 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21776630 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21776629 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21776627 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005754 |
BC013803.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005754 |
NM_026968.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005754 |
AK007599.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005753 |
BC013803.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005753 |
NM_026968.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2005753 |
AK007599.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
467080 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931531 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
429077 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
604819 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
385566 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
637538 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
544543 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
450907 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
125561 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
833380 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
613998 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
541304 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
354078 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
612957 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696083 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
597729 |
AK015788.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
597729 |
XM_136059.4 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
138796 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
114391 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P135756 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P125067 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2005753 |
BC013803.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2005753 |
NM_026968.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2005753 |
AK007599.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2073163 |
AK079707.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2005754 |
BC013803.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2005754 |
NM_026968.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2005754 |
AK007599.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776606 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776630 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776615 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776625 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776607 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776618 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776609 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776627 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776622 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776611 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776619 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776616 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776612 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776637 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776632 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776636 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776626 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776624 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776604 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776634 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21776602 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P135756 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04754577 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P125067 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P440107 |
AK169993.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P440107 |
BC013803.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P440107 |
BC030375.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P440107 |
NM_026968.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P440107 |
AK007599.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P135756 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P125067 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02123488 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01897221 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02123487 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01828851 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21776629 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21776627 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21776630 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21776628 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21776626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776698 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776822 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776810 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776619 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776785 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776745 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776777 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776818 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776692 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776782 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776781 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776751 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776797 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776673 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776752 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776599 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776711 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776762 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776615 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776681 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776793 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776754 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776824 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776756 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776684 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776798 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776738 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776817 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776763 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776778 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776678 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776789 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776592 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776764 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776636 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776821 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21776736 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01778732 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01702175 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776736 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01702183 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776709 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01694034 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02110965 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776643 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776642 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776616 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01539204 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01460888 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776609 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01391361 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01150882 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776649 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01432081 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776632 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776733 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776612 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01694037 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01432084 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01843736 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776663 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776599 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776740 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776673 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776745 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776756 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776737 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01694043 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01694048 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01702177 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776666 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776647 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02123483 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01981676 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01590437 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01722119 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776604 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01978599 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01590931 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776598 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01694049 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776625 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02127670 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776731 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776668 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776757 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776748 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776707 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776640 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01171150 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01828851 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776716 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776648 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776655 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01694041 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776597 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01678449 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776713 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02067554 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01827466 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776658 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02123485 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776618 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776611 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776636 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01828850 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02218798 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776738 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776729 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01432085 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776626 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776664 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21776615 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01150887 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02110967 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01694047 |
- |
- |