Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028877.1 | NP_083153.1 | 4 - 5 | 329 | 64 | |
GenBank mRNA | AK014493.1 | - | 4 - 5 | 333 | 64 | |
BC113196.1 | - | 4 - 5 | 332 | 64 | ||
BC113802.1 | - | 4 - 5 | 327 | 64 | ||
EF208034.1 | - | 4 - 5 | 329 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |