Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024617.3 | NP_001019788.1 | 4 - 5 | 566 | 60 | |
NM_001297591.1 | NP_001284520.1 | 3 - 4 | 492 | 60 | ||
NM_001297593.1 | NP_001284522.1 | 3 - 4 | 492 | 60 | ||
NM_001297596.1 | NP_001284525.1 | 3 - 4 | 492 | 60 | ||
GenBank mRNA | AK141502.1 | - | 5 - 6 | 489 | 60 | |
AK172564.1 | - | 4 - 5 | 581 | 60 | ||
AK172601.1 | - | 4 - 5 | 581 | 60 | ||
BC064813.1 | - | 2 - 3 | 268 | 6826283 | 60 | |
DQ296611.1 | - | 3 - 4 | 492 | 60 | ||
DQ296612.1 | - | 3 - 4 | 492 | 60 | ||
DQ296613.1 | - | 3 - 4 | 492 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |