Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081981.2 | NP_001075450.1 | 1 - 2 | 1013 | 57 | |
NM_001297601.1 | NP_001284530.1 | 1 - 2 | 1013 | 57 | ||
GenBank mRNA | AK158568.1 | - | 1 - 2 | 1016 | 57 | |
BC090626.1 | - | 1 - 2 | 40 | 57 | ||
CX221451.1 | - | 1 - 2 | 117 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |