Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037877.3 | NP_001032966.1 | 1 - 2 | 353 | 67 | |
NM_001037878.3 | NP_001032967.1 | 1 - 2 | 353 | 67 | ||
NM_001286362.1 | NP_001273291.1 | 1 - 2 | 353 | 67 | ||
NM_001286363.1 | NP_001273292.1 | 1 - 2 | 353 | 67 | ||
NM_025804.3 | NP_080080.2 | 1 - 2 | 353 | 67 | ||
GenBank mRNA | AK003180.1 | - | 1 - 2 | 241 | 67 | |
AK013754.1 | - | 1 - 2 | 42 | 67 | ||
AK045397.1 | - | 1 - 2 | 281 | 67 | ||
AK050274.1 | - | 1 - 2 | 281 | 67 | ||
AK075728.1 | - | 1 - 2 | 207 | 67 | ||
AK088360.1 | - | 1 - 2 | 241 | 67 | ||
AK089964.1 | - | 1 - 2 | 242 | 67 | ||
AK122430.1 | - | 1 - 2 | 208 | 67 | ||
AK150232.1 | - | 1 - 2 | 240 | 67 | ||
AK150918.1 | - | 1 - 2 | 234 | 67 | ||
AK150942.1 | - | 1 - 2 | 227 | 67 | ||
AK151143.1 | - | 1 - 2 | 227 | 67 | ||
AK151558.1 | - | 1 - 2 | 234 | 67 | ||
AK152178.1 | - | 1 - 2 | 227 | 67 | ||
AK152494.1 | - | 1 - 2 | 227 | 67 | ||
AK161627.1 | - | 1 - 2 | 229 | 67 | ||
AK167837.1 | - | 1 - 2 | 227 | 67 | ||
AK168167.1 | - | 1 - 2 | 227 | 67 | ||
BC016130.1 | - | 1 - 2 | 217 | 67 | ||
BC025071.1 | - | 1 - 2 | 214 | 3498003 | 67 | |
BC046768.1 | - | 1 - 2 | 311 | 5707312 | 67 | |
EU624323.1 | - | 1 - 2 | 191 | 67 | ||
U94988.1 | - | 1 - 2 | 214 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |