Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chtf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145412.3 | NP_663387.3 | 1 - 2 | 97 | 102 | |
GenBank mRNA | AK030924.1 | - | 1 - 2 | 96 | 102 | |
AK088257.1 | - | 1 - 2 | 106 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |