Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272052.1 | NP_001258981.1 | 6 - 7 | 757 | 57 | |
NM_007398.4 | NP_031424.1 | 6 - 7 | 757 | 57 | ||
GenBank mRNA | AF483480.1 | - | 6 - 7 | 609 | 57 | |
AF483481.1 | - | 6 - 7 | 609 | 57 | ||
AK075899.1 | - | 6 - 7 | 696 | 57 | ||
AK167489.1 | - | 6 - 7 | 696 | 57 | ||
BC002075.1 | - | 6 - 7 | 707 | 3484465 | 57 | |
CT010224.1 | - | 6 - 7 | 609 | 57 | ||
CT010255.1 | - | 6 - 7 | 609 | 57 | ||
M10319.1 | - | 6 - 7 | 679 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |