Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272052.1 | NP_001258981.1 | 9 - 10 | 994 | 69 | |
NM_007398.4 | NP_031424.1 | 9 - 10 | 994 | 69 | ||
GenBank mRNA | AF483480.1 | - | 9 - 10 | 846 | 69 | |
AF483481.1 | - | 9 - 10 | 846 | 69 | ||
AK075899.1 | - | 9 - 10 | 933 | 69 | ||
AK167489.1 | - | 9 - 10 | 933 | 69 | ||
BC002075.1 | - | 9 - 10 | 944 | 3484465 | 69 | |
CT010224.1 | - | 9 - 10 | 846 | 69 | ||
CT010255.1 | - | 9 - 10 | 846 | 69 | ||
M10319.1 | - | 9 - 10 | 916 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |